Alfa Aesar™ 3-Fluoro-1,2-dimethyl-6-nitrobenzene, 98%

Manufacturer: Alfa Aesar™ H3255706

Quantity 5g

 View more versions of this product

Catalog No. AAH3255706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1736-87-4
    Molecular Formula C8H8FNO2
    Formula Weight 169.16
    MDL Number MFCD05663712
    UN Number UN2811
    Synonym 3-Fluoro-6-nitro-o-xylene
    Chemical Name or Material 3-Fluoro-1,2-dimethyl-6-nitrobenzene

    Specifications

    Formula Weight 169.16
    Flash Point >110°C (230°F)
    Molecular Formula C8H8FNO2
    MDL Number MFCD05663712
    CAS 1736-87-4
    UN Number UN2811
    Synonym 3-Fluoro-6-nitro-o-xylene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Odor Characteristic
    Melting Point 39° to 42°C
    Quantity 5g
    Chemical Name or Material 3-Fluoro-1,2-dimethyl-6-nitrobenzene
    Assay 98%

    GHS H Statement
    H311-H302-H332-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit