Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-2-formylbenzeneboronic acid, 95%

Click to view available options
Quantity:
1g
5g
25g
871126-15-7
C7H6BFO3
167.93
MFCD10697421
GVHWLCYABLWGIR-UHFFFAOYSA-N
3-fluoro-2-formylphenyl boronic acid,3-fluoro-2-formylbenzeneboronic acid,boronic acid, 3-fluoro-2-formylphenyl,acmc-209qf3,2-formyl-3-fluorophenylboranic acid,3-fluoro-2-formyl-phenyl boronic acid,3-fluoro-2-formylphenylboronic acid,boronic acid, b-3-fluoro-2-formylphenyl,contains varying amounts of anhydride
53412038
(3-fluoro-2-formylphenyl)boronic acid
B(C1=C(C(=CC=C1)F)C=O)(O)O
871126-15-7
C7H6BFO3
167.93
MFCD10697421
GVHWLCYABLWGIR-UHFFFAOYSA-N
3-fluoro-2-formylphenyl boronic acid,3-fluoro-2-formylbenzeneboronic acid,boronic acid, 3-fluoro-2-formylphenyl,acmc-209qf3,2-formyl-3-fluorophenylboranic acid,3-fluoro-2-formyl-phenyl boronic acid,3-fluoro-2-formylphenylboronic acid,boronic acid, b-3-fluoro-2-formylphenyl,contains varying amounts of anhydride
53412038
(3-fluoro-2-formylphenyl)boronic acid
B(C1=C(C(=CC=C1)F)C=O)(O)O
Applications
Undergoes unprecedented tautomeric rearrangement in solution.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Melting Point 123°C to 127°C
Quantity 5g
Sensitivity Light Sensitive
UN Number UN3261
Formula Weight 167.93
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-Fluoro-2-formylbenzeneboronic acid

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C