Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-2-methoxy-6-methylpyridine, 97%

Click to view available options
Quantity:
1g
5g
375368-80-2
C7H8FNO
141.145
MFCD03095083
DAQMWRIPXIPMAA-UHFFFAOYSA-N
3-fluoro-2-methoxy-6-picoline,2-methoxy-3-fluoro-6-methylpyridine,3-fluoro-2-methoxy-6-methyl-pyridine,pubchem6585,ksc914i9l,abbypharma ap-15-5217,pyridine, 3-fluoro-2-methoxy-6-methyl,pyridine, 3-fluoro-2-methoxy-6-methyl-9ci
24820319
3-fluoro-2-methoxy-6-methylpyridine
CC1=NC(=C(C=C1)F)OC
375368-80-2
C7H8FNO
141.145
MFCD03095083
DAQMWRIPXIPMAA-UHFFFAOYSA-N
3-fluoro-2-methoxy-6-picoline,2-methoxy-3-fluoro-6-methylpyridine,3-fluoro-2-methoxy-6-methyl-pyridine,pubchem6585,ksc914i9l,abbypharma ap-15-5217,pyridine, 3-fluoro-2-methoxy-6-methyl,pyridine, 3-fluoro-2-methoxy-6-methyl-9ci
24820319
3-fluoro-2-methoxy-6-methylpyridine
CC1=NC(=C(C=C1)F)OC
Quantity 1g
Color Colorless to Yellow
Formula Weight 141.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Fluoro-2-methoxy-6-methylpyridine