Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-2-methylaniline, 98+%

Click to view available options
Quantity:
5g
25g
443-86-7
C7H8FN
125.146
MFCD00007760
SLDLVGFPFFLYBM-UHFFFAOYSA-N
3-fluoro-o-toluidine,2-fluoro-6-aminotoluene,2-amino-6-fluorotoluene,6-amino-2-fluorotoluene,3-fluoro-2-methyl-aniline,benzenamine, 3-fluoro-2-methyl,3-fluoro-2-methylphenylamine,2-methyl-3-fluoroaniline,hydrochloride,3-fluor-2-methylanilin
285553
3-fluoro-2-methylaniline
CC1=C(C=CC=C1F)N
443-86-7
C7H8FN
125.146
MFCD00007760
SLDLVGFPFFLYBM-UHFFFAOYSA-N
3-fluoro-o-toluidine,2-fluoro-6-aminotoluene,2-amino-6-fluorotoluene,6-amino-2-fluorotoluene,3-fluoro-2-methyl-aniline,benzenamine, 3-fluoro-2-methyl,3-fluoro-2-methylphenylamine,2-methyl-3-fluoroaniline,hydrochloride,3-fluor-2-methylanilin
285553
3-fluoro-2-methylaniline
CC1=C(C=CC=C1F)N
Flash Point 86°C (186°F)
Boiling Point 205°C
Melting Point 7°C
Odor Characteristic
Quantity 25g
UN Number UN2810
Formula Weight 125.15
Percent Purity ≥98%
Chemical Name or Material 3-Fluoro-2-methylaniline

GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P260-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.