Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-2-methylbenzaldehyde, 98%

Click to view available options
Quantity:
1g
5g
147624-13-3
C8H7FO
138.141
MFCD00075256
HSUCYMJBFHBMTB-UHFFFAOYSA-N
2-methyl-3-fluorobenzaldehyde,3-fluoro-2-methyl-benzaldehyde,benzaldehyde, 3-fluoro-2-methyl,pubchem1453,acmc-1cfbd,intermediates-zcf02128,ksc494i0h,attercop-chm at111499,3-fluoro-2-methylbenzaldehyde
2779235
3-fluoro-2-methylbenzaldehyde
CC1=C(C=CC=C1F)C=O
147624-13-3
C8H7FO
138.141
MFCD00075256
HSUCYMJBFHBMTB-UHFFFAOYSA-N
2-methyl-3-fluorobenzaldehyde,3-fluoro-2-methyl-benzaldehyde,benzaldehyde, 3-fluoro-2-methyl,pubchem1453,acmc-1cfbd,intermediates-zcf02128,ksc494i0h,attercop-chm at111499,3-fluoro-2-methylbenzaldehyde
2779235
3-fluoro-2-methylbenzaldehyde
CC1=C(C=CC=C1F)C=O
Flash Point 81°C (178°F)
Refractive Index 1.526
Density 1.16 g/mL
Quantity 5g
Color Yellow
Boiling Point 92°C to 93°C (20mmHg)
Beilstein 6321735
Formula Weight 138.14
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Fluoro-2-methylbenzaldehyde
Show More Show Less

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold