Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-2-methylpyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
15931-15-4
C6H6FN
111.119
MFCD08705710
IRIHSZDBTACXCT-UHFFFAOYSA-N
3-fluoro-2-picoline,pyridine, 3-fluoro-2-methyl,2-methyl-3-fluoropyridine,3-fluoro-2-methyl pyridine,3-fluoro-2-methyl-pyridine,fluoropicoline,pubchem14381,acmc-209xci,ksc530a3b,pyridine,3-fluoro-2-methyl
21613172
3-fluoro-2-methylpyridine
CC1=C(C=CC=N1)F
15931-15-4
C6H6FN
111.119
MFCD08705710
IRIHSZDBTACXCT-UHFFFAOYSA-N
3-fluoro-2-picoline,pyridine, 3-fluoro-2-methyl,2-methyl-3-fluoropyridine,3-fluoro-2-methyl pyridine,3-fluoro-2-methyl-pyridine,fluoropicoline,pubchem14381,acmc-209xci,ksc530a3b,pyridine,3-fluoro-2-methyl
21613172
3-fluoro-2-methylpyridine
CC1=C(C=CC=N1)F
Applications
3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
Color Colorless to Yellow
Beilstein 6497832
Formula Weight 111.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Fluoro-2-methylpyridine

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger