Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-2-nitrophenol, 97%

Click to view available options
Quantity:
1g
5g
385-01-3
C6H4FNO3
157.1
MFCD07368753
GAWNBKUTBVLIPL-UHFFFAOYSA-N
3-fluoro-2-nitro-phenol,2-nitro-3-fluorophenol,phenol, 3-fluoro-2-nitro,3-fluoronitro-phenol,pubchem4124,acmc-209j0i,ksc497m1h
2782777
3-fluoro-2-nitrophenol
C1=CC(=C(C(=C1)F)[N+](=O)[O-])O
385-01-3
C6H4FNO3
157.1
MFCD07368753
GAWNBKUTBVLIPL-UHFFFAOYSA-N
3-fluoro-2-nitro-phenol,2-nitro-3-fluorophenol,phenol, 3-fluoro-2-nitro,3-fluoronitro-phenol,pubchem4124,acmc-209j0i,ksc497m1h
2782777
3-fluoro-2-nitrophenol
C1=CC(=C(C(=C1)F)[N+](=O)[O-])O
Quantity 1g
Formula Weight 157.1
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Fluoro-2-nitrophenol

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures