Learn More
Alfa Aesar™ 3-Fluoro-2-(trifluoromethyl)benzylamine, 97%
Manufacturer:  Alfa Aesar™ H32210MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH32210MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
771581-62-5
193.15
UN2735
C8H7F4N
MFCD06212983
3-Fluoro-2-(trifluoromethyl)benzylamine
Specifications
250mg
C8H7F4N
MFCD06212983
3-Fluoro-2-(trifluoromethyl)benzylamine
771581-62-5
193.15
UN2735
97%
Safety and Handling
SDS & Safety Documents
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.