Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-formylphenylboronic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
248270-25-9
C7H6BFO3
167.93
NZNRMUVHUVCIBR-UHFFFAOYSA-N
3-fluoro-4-formylbenzeneboronic acid,3-fluoro-4-formylphenyl boronic acid,3-fluoro-4-formylphenboronic acid,boronic acid, 3-fluoro-4-formylphenyl,pubchem1847,acmc-1cga0,ksc494k4h,4-borono-3-fluorobenzaldehyde,3-fluoro-4-formyl-phenyl boronic acid,3-fluoranyl-4-methanoyl-phenyl boronic acid
2778654
(3-fluoro-4-formylphenyl)boronic acid
B(C1=CC(=C(C=C1)C=O)F)(O)O
248270-25-9
C7H6BFO3
167.93
NZNRMUVHUVCIBR-UHFFFAOYSA-N
3-fluoro-4-formylbenzeneboronic acid,3-fluoro-4-formylphenyl boronic acid,3-fluoro-4-formylphenboronic acid,boronic acid, 3-fluoro-4-formylphenyl,pubchem1847,acmc-1cga0,ksc494k4h,4-borono-3-fluorobenzaldehyde,3-fluoro-4-formyl-phenyl boronic acid,3-fluoranyl-4-methanoyl-phenyl boronic acid
2778654
(3-fluoro-4-formylphenyl)boronic acid
B(C1=CC(=C(C=C1)C=O)F)(O)O
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Formula Weight 167.93
Percent Purity 95%
Chemical Name or Material 3-Fluoro-4-formylphenylboronic acid

GHS H Statement
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning