Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-iodopyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
22282-75-3
C5H3FIN
222.98
MSLUTDVOKZOXTA-UHFFFAOYSA-N
3-fluoro-4-iodo-pyridine,4-iodo-3-fluoropyridine,pyridine, 3-fluoro-4-iodo,pubchem9249,acmc-1ccsk,pyridine,3-fluoro-4-iodo,3-fluoro-4-iodopyridine,3-fluoro-4-iodopyridine 1g
10998636
3-fluoro-4-iodopyridine
C1=CN=CC(=C1I)F
22282-75-3
C5H3FIN
222.98
MSLUTDVOKZOXTA-UHFFFAOYSA-N
3-fluoro-4-iodo-pyridine,4-iodo-3-fluoropyridine,pyridine, 3-fluoro-4-iodo,pubchem9249,acmc-1ccsk,pyridine,3-fluoro-4-iodo,3-fluoro-4-iodopyridine,3-fluoro-4-iodopyridine 1g
10998636
3-fluoro-4-iodopyridine
C1=CN=CC(=C1I)F
Packaging Glass Bottle
Infrared Spectrum Authentic
Color Cream to Beige
Quantity 1g
Melting Point 189.4°C to 190.4°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 222.98
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-iodopyridine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning