Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-methoxyaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
366-99-4
C7H8FNO
141.15
MFCD00075040
LJWAPDSCYTZUJU-UHFFFAOYSA-N
3-fluoro-4-anisidine,4-amino-2-fluoroanisole,3-fluoro-p-anisidine,3-fluoro-4-methoxy-phenylamine,benzenamine, 3-fluoro-4-methoxy,3-fluoro-4-methoxy-aniline,3-fluoro-4-methoxyphenylamine,2-fluoro-4-aminoanisole,3-fluoro-4-methoxy-anilin,3-fluoro-4-methoxy-benzenamine
581110
3-fluoro-4-methoxyaniline
COC1=C(C=C(C=C1)N)F
366-99-4
C7H8FNO
141.15
MFCD00075040
LJWAPDSCYTZUJU-UHFFFAOYSA-N
3-fluoro-4-anisidine,4-amino-2-fluoroanisole,3-fluoro-p-anisidine,3-fluoro-4-methoxy-phenylamine,benzenamine, 3-fluoro-4-methoxy,3-fluoro-4-methoxy-aniline,3-fluoro-4-methoxyphenylamine,2-fluoro-4-aminoanisole,3-fluoro-4-methoxy-anilin,3-fluoro-4-methoxy-benzenamine
581110
3-fluoro-4-methoxyaniline
COC1=C(C=C(C=C1)N)F
Boiling Point 135°C (18.0mmHg)
Melting Point 81°C to 84°C
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige to Brown-Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula FC6H3(NH2)OCH3
Formula Weight 141.15
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 3-Fluoro-4-methoxyaniline
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact

GHS Signal Word: Warning