Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-4-methoxybenzoic acid, 98%

Click to view available options
Quantity:
1g
5g
403-20-3
C8H7FO3
170.139
MFCD00060675
HYNNNQDQEORWEU-UHFFFAOYSA-N
3-fluoro-4-methoxy-benzoic acid,3-fluoro-p-anisic acid,3-fluoro-4-methoxy benzoic acid,3-fluoro-4-methoxybenzoicacid,pubchem1393,opera_id_903,acmc-209jd1,ksc494s2f,3-fluoro-4-methoxy benzoicacid,rarechem al bo 0870
2733401
3-fluoro-4-methoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)F
403-20-3
C8H7FO3
170.139
MFCD00060675
HYNNNQDQEORWEU-UHFFFAOYSA-N
3-fluoro-4-methoxy-benzoic acid,3-fluoro-p-anisic acid,3-fluoro-4-methoxy benzoic acid,3-fluoro-4-methoxybenzoicacid,pubchem1393,opera_id_903,acmc-209jd1,ksc494s2f,3-fluoro-4-methoxy benzoicacid,rarechem al bo 0870
2733401
3-fluoro-4-methoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)F
Quantity 1g
Melting Point 209°C to 213°C
Beilstein 2087583
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures