Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-4-methoxybenzoic acid, 98%

Click to view available options
Quantity:
1g
5g
403-20-3
C8H7FO3
170.139
MFCD00060675
HYNNNQDQEORWEU-UHFFFAOYSA-N
3-fluoro-4-methoxy-benzoic acid,3-fluoro-p-anisic acid,3-fluoro-4-methoxy benzoic acid,3-fluoro-4-methoxybenzoicacid,pubchem1393,opera_id_903,acmc-209jd1,ksc494s2f,3-fluoro-4-methoxy benzoicacid,rarechem al bo 0870
2733401
3-fluoro-4-methoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)F
403-20-3
C8H7FO3
170.139
MFCD00060675
HYNNNQDQEORWEU-UHFFFAOYSA-N
3-fluoro-4-methoxy-benzoic acid,3-fluoro-p-anisic acid,3-fluoro-4-methoxy benzoic acid,3-fluoro-4-methoxybenzoicacid,pubchem1393,opera_id_903,acmc-209jd1,ksc494s2f,3-fluoro-4-methoxy benzoicacid,rarechem al bo 0870
2733401
3-fluoro-4-methoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)F
Quantity 5g
Melting Point 209°C to 213°C
Beilstein 2087583
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning