Alfa Aesar™ 3-Fluoro-4-methoxybenzylamine, 97%

Manufacturer: Alfa Aesar™ H2660703

Quantity 1g

 View more versions of this product

Catalog No. AAH2660703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 123652-95-9
    Molecular Formula C8H10FNO
    Formula Weight 155.17
    MDL Number MFCD04116360
    UN Number UN2735
    Chemical Name or Material 3-Fluoro-4-methoxybenzylamine

    Specifications

    Formula Weight 155.17
    Molecular Formula C8H10FNO
    MDL Number MFCD04116360
    CAS 123652-95-9
    UN Number UN2735
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Quantity 1g
    Chemical Name or Material 3-Fluoro-4-methoxybenzylamine
    Assay 97%

    Description

    Applications
    3-Fluoro-4-methoxybenzylamine is used as an intermediate in organic synthesis.

    Solubility
    Immiscible with water.

    Notes
    Air sensitive. Store in a cool place. Incompatible with oxidizing agents, bases, acids and carbon dioxide.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit