Alfa Aesar™ 3-Fluoro-4-methyl-DL-phenylalanine, 97%

Manufacturer: Alfa Aesar™ H32194MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32194MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C10H12FNO2
    Formula Weight 197.21
    MDL Number MFCD09832258
    Chemical Name or Material 3-Fluoro-4-methyl-DL-phenylalanine

    Specifications

    Formula Weight 197.21
    Melting Point ≈368°C
    Molecular Formula C10H12FNO2
    MDL Number MFCD09832258
    Quantity 250mg
    Chemical Name or Material 3-Fluoro-4-methyl-DL-phenylalanine
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit