Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3'-Fluoro-4'-methylacetophenone, 97+%

Click to view available options
Quantity:
1g
5g
42444-14-4
C9H9FO
152.168
MFCD01631411
XAMNMGLEJNLUIH-UHFFFAOYSA-N
3'-fluoro-4'-methylacetophenone,1-3-fluoro-4-methylphenyl ethanone,1-3-fluoro-4-methylphenyl ethan-1-one,3-fluoro-4-methylacetophenone,1-acetyl-3-fluoro-4-methylbenzene,4-acetyl-2-fluorotoluene
2774569
1-(3-fluoro-4-methylphenyl)ethanone
CC1=C(C=C(C=C1)C(=O)C)F
42444-14-4
C9H9FO
152.168
MFCD01631411
XAMNMGLEJNLUIH-UHFFFAOYSA-N
3'-fluoro-4'-methylacetophenone,1-3-fluoro-4-methylphenyl ethanone,1-3-fluoro-4-methylphenyl ethan-1-one,3-fluoro-4-methylacetophenone,1-acetyl-3-fluoro-4-methylbenzene,4-acetyl-2-fluorotoluene
2774569
1-(3-fluoro-4-methylphenyl)ethanone
CC1=C(C=C(C=C1)C(=O)C)F
Melting Point 31°C to 34°C
Quantity 1g
Formula Weight 152.17
Percent Purity ≥97%
Chemical Name or Material 3'-Fluoro-4'-methylacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning