Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-4-methylbenzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
177756-62-6
C8H7FO
138.141
MFCD01631532
UFPBMVRONDLOGK-UHFFFAOYSA-N
3-fluoro-p-tolualdehyde,3-fluoro-4-methyl-benzaldehyde,benzaldehyde, 3-fluoro-4-methyl-9ci,pubchem1454,acmc-1c3kx,ksc494i5j,3-fluoranyl-4-methyl-benzaldehyde,3-fluoro-4-methylbenzaldehyde,benzaldehyde, 3-fluoro-4-methyl
2774574
3-fluoro-4-methylbenzaldehyde
CC1=C(C=C(C=C1)C=O)F
177756-62-6
C8H7FO
138.141
MFCD01631532
UFPBMVRONDLOGK-UHFFFAOYSA-N
3-fluoro-p-tolualdehyde,3-fluoro-4-methyl-benzaldehyde,benzaldehyde, 3-fluoro-4-methyl-9ci,pubchem1454,acmc-1c3kx,ksc494i5j,3-fluoranyl-4-methyl-benzaldehyde,3-fluoro-4-methylbenzaldehyde,benzaldehyde, 3-fluoro-4-methyl
2774574
3-fluoro-4-methylbenzaldehyde
CC1=C(C=C(C=C1)C=O)F
Quantity 5g
Boiling Point 206°C
Beilstein 7699714
Formula Weight 138.14
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylbenzaldehyde

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning