Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-4-methylbenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
168267-99-0
C7H8BFO2
153.947
MFCD02683115
WPVBHUUZDFUIJA-UHFFFAOYSA-N
3-fluoro-4-methylbenzeneboronic acid,3-fluoro-4-methylphenyl boronic acid,3-fluoro-p-tolylboronic acid,3-fluoro-4-methylphenyl-boronic acid,boronic acid, 3-fluoro-4-methylphenyl,pubchem5124,3-fluoro-4-methyl-phenyl boronic acid,acmc-209dxx,ksc173s0n
2782674
(3-fluoro-4-methylphenyl)boronic acid
B(C1=CC(=C(C=C1)C)F)(O)O
168267-99-0
C7H8BFO2
153.947
MFCD02683115
WPVBHUUZDFUIJA-UHFFFAOYSA-N
3-fluoro-4-methylbenzeneboronic acid,3-fluoro-4-methylphenyl boronic acid,3-fluoro-p-tolylboronic acid,3-fluoro-4-methylphenyl-boronic acid,boronic acid, 3-fluoro-4-methylphenyl,pubchem5124,3-fluoro-4-methyl-phenyl boronic acid,acmc-209dxx,ksc173s0n
2782674
(3-fluoro-4-methylphenyl)boronic acid
B(C1=CC(=C(C=C1)C)F)(O)O
Melting Point 234°C
Quantity 1g
Beilstein 8544594
Formula Weight 153.95
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning