Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-methylbenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
2.5g
Packaging:
Glass bottle
350-28-7
C8H7FO2
154.14
XUQCONCMPCVUDM-UHFFFAOYSA-N
3-fluoro-p-toluic acid,3-fluoro-4-methylbenzoicacid,3-fluoro-4-methyl-benzoic acid,benzoic acid, 3-fluoro-4-methyl,pubchem1330,acmc-209ich,4-carboxy-2-fluorotoluene,3-fluoro4-methylbenzoic acid,ksc223k8t,rarechem al bo 0761
67687
3-fluoro-4-methylbenzoic acid
CC1=C(C=C(C=C1)C(=O)O)F
350-28-7
C8H7FO2
154.14
XUQCONCMPCVUDM-UHFFFAOYSA-N
3-fluoro-p-toluic acid,3-fluoro-4-methylbenzoicacid,3-fluoro-4-methyl-benzoic acid,benzoic acid, 3-fluoro-4-methyl,pubchem1330,acmc-209ich,4-carboxy-2-fluorotoluene,3-fluoro4-methylbenzoic acid,ksc223k8t,rarechem al bo 0761
67687
3-fluoro-4-methylbenzoic acid
CC1=C(C=C(C=C1)C(=O)O)F
Quantity 2.5g
Infrared Spectrum Authentic
Color Beige-Gray to White
Melting Point 170.0°C to 175.0°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H3(CH3)CO2H
Beilstein 09, 497
Formula Weight 154.14
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylbenzoic acid, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.