Alfa Aesar™ 3-Fluoro-4-methylbenzoyl chloride, 97%

Manufacturer: Alfa Aesar™ B2284606

Quantity 5g

 View more versions of this product

Catalog No. AAB2284606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 59189-97-8
    Molecular Formula C8H6ClFO
    Formula Weight 172.59
    MDL Number MFCD00671774
    UN Number UN3265
    Beilstein 7757348
    Synonym 3-Fluoro-p-toluoyl chloride
    Chemical Name or Material 3-Fluoro-4-methylbenzoyl chloride

    Specifications

    Formula Weight 172.59
    Molecular Formula C8H6ClFO
    Quantity 5g
    MDL Number MFCD00671774
    UN Number UN3265
    Beilstein 7757348
    Synonym 3-Fluoro-p-toluoyl chloride
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 59189-97-8
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 3-Fluoro-4-methylbenzoyl chloride
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit