Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-methylbenzoyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
59189-97-8
C8H6ClFO
172.583
MFCD00671774
DPWCZSXNEGNALT-UHFFFAOYSA-N
3-fluoro-4-methyl-benzoyl chloride,3-fluoro-4-methylbenzoylchloride,acmc-20an3p,3-fluoro-4-methyl-benzoylchloride,3-fluoro-4-methyl benzoyl chloride,benzoyl chloride,3-fluoro-4-methyl,3-fluoranyl-4-methyl-benzoyl chloride,benzoyl chloride, 3-fluoro-4-methyl,3-fluoro-4-methylbenzoyl chloride,benzoyl chloride, 3-fluoro-4-methyl-9ci
2734875
3-fluoro-4-methylbenzoyl chloride
CC1=C(C=C(C=C1)C(=O)Cl)F
59189-97-8
C8H6ClFO
172.583
MFCD00671774
DPWCZSXNEGNALT-UHFFFAOYSA-N
3-fluoro-4-methyl-benzoyl chloride,3-fluoro-4-methylbenzoylchloride,acmc-20an3p,3-fluoro-4-methyl-benzoylchloride,3-fluoro-4-methyl benzoyl chloride,benzoyl chloride,3-fluoro-4-methyl,3-fluoranyl-4-methyl-benzoyl chloride,benzoyl chloride, 3-fluoro-4-methyl,3-fluoro-4-methylbenzoyl chloride,benzoyl chloride, 3-fluoro-4-methyl-9ci
2734875
3-fluoro-4-methylbenzoyl chloride
CC1=C(C=C(C=C1)C(=O)Cl)F
Infrared Spectrum Authentic
Refractive Index 1.5320 to 1.5340 (20°C, 589nm)
Quantity 5g
Color Yellow
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C6H3(F)C(O)Cl
Formula Weight 172.59
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-Fluoro-4-methylbenzoyl chloride
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger