Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-methylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
50mL
Packaging:
AcroSeal™ Glass Bottle
185077-02-5,109-99-9
C7H6BrFMg
213.33
MFCD01311470
QLGSDGDHUROFKU-UHFFFAOYSA-M
3-fluoro-4-methylphenylmagnesium bromide,bromo 3-fluoro-4-methylphenyl magnesium,3-fluoro-4-methylphenylmagnesium bromide 0.5m solution in thf,fgtairviejuzjo-uhfffaoysa-m,3-fluoro-4-methylphenyl magnesiumbromide,magnesium,bromo 3-fluoro-4-methylphenyl,3-fluoro-4-methylphenylmagnesium bromide, 0.50 m in 2-methf,3-fluoro-4-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran,3-fluoro-4-methylphenylmagnesium bromide solution, 0.5 m in thf
2778411
magnesium;1-fluoro-2-methylbenzene-5-ide;bromide
CC1=CC=[C-]C=C1F.[Mg+2].[Br-]
185077-02-5,109-99-9
C7H6BrFMg
213.33
MFCD01311470
QLGSDGDHUROFKU-UHFFFAOYSA-M
3-fluoro-4-methylphenylmagnesium bromide,bromo 3-fluoro-4-methylphenyl magnesium,3-fluoro-4-methylphenylmagnesium bromide 0.5m solution in thf,fgtairviejuzjo-uhfffaoysa-m,3-fluoro-4-methylphenyl magnesiumbromide,magnesium,bromo 3-fluoro-4-methylphenyl,3-fluoro-4-methylphenylmagnesium bromide, 0.50 m in 2-methf,3-fluoro-4-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran,3-fluoro-4-methylphenylmagnesium bromide solution, 0.5 m in thf
2778411
magnesium;1-fluoro-2-methylbenzene-5-ide;bromide
CC1=CC=[C-]C=C1F.[Mg+2].[Br-]
Specific Gravity 0.953
Density 0.9530g/mL
Flash Point −17°C
Color Yellow
Packaging AcroSeal™ Glass Bottle
Quantity 50mL
CAS Min % 8.95,86.57
CAS Max % 13.43,91.05
Linear Formula F(CH3)C6H3MgBr
Solubility Information Solubility in water: reacts
Formula Weight 213.33
Physical Form Solution
Chemical Name or Material 3-Fluoro-4-methylphenylmagnesium bromide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger