Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-4-methylpyridine, 95%

Click to view available options
Quantity:
1g
5g
399-88-2
C6H6FN
111.119
MFCD04114226
WZMOEPZZTTWDIA-UHFFFAOYSA-N
3-fluoro-4-picoline,4-methyl-3-fluoropyridine,pyridine, 3-fluoro-4-methyl,zlchem 994,pubchem9251,acmc-1agea,3-fluoro-4-methyl pyridine,3-fluoro-4-methyl-pyridine,3-fluoro-4-methylpyridine?,ksc925s4f
345367
3-fluoro-4-methylpyridine
CC1=C(C=NC=C1)F
399-88-2
C6H6FN
111.119
MFCD04114226
WZMOEPZZTTWDIA-UHFFFAOYSA-N
3-fluoro-4-picoline,4-methyl-3-fluoropyridine,pyridine, 3-fluoro-4-methyl,zlchem 994,pubchem9251,acmc-1agea,3-fluoro-4-methyl pyridine,3-fluoro-4-methyl-pyridine,3-fluoro-4-methylpyridine?,ksc925s4f
345367
3-fluoro-4-methylpyridine
CC1=C(C=NC=C1)F
Density 1.08g/mL
Color Colorless to Yellow
Quantity 5g
Boiling Point 136°C (748mmHg)
Refractive Index 1.4795
UN Number UN1993
Beilstein 107788
Formula Weight 111.12
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 3-Fluoro-4-methylpyridine
Show More Show Less

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.