Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-nitrobenzaldehyde, TCI America™

Click to view available options
Quantity:
1g
5g
160538-51-2
C7H4FNO3
169.111
MFCD00968940
BWUIGISQVCIQBT-UHFFFAOYSA-N
3808120
3-fluoro-4-nitrobenzaldehyde
C1=CC(=C(C=C1C=O)F)[N+](=O)[O-]
160538-51-2
C7H4FNO3
169.111
MFCD00968940
BWUIGISQVCIQBT-UHFFFAOYSA-N
3808120
3-fluoro-4-nitrobenzaldehyde
C1=CC(=C(C=C1C=O)F)[N+](=O)[O-]
Melting Point 55°C
Color Undesignated
Quantity 5g
Formula Weight 169.11
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material 3-Fluoro-4-nitrobenzaldehyde