Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-nitrobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7031045
Click to view available options
Quantity:
1 g
86156-93-6
C6H3ClFNO4S
239.60
MFCD03412240
AAWFUSFNPBRQDE-UHFFFAOYSA-N
3-fluoro-4-nitrobenzene-1-sulfonyl chloride, 3-fluoro-4-nitro-benzenesulfonyl chloride, 3-fluoro-4-nitrobenzenesulfonylchloride, 3-fluro-4-nitro-benzensulfonylchloride, 3-fluoro-4-nitrophenylsulfonyl chloride, benzenesulfonylchloride, 3-fluoro-4-nitro, benzenesulfonyl chloride, 3-fluoro-4-nitro, 3-fluoro-4-nitro-benzensulfonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-nitro-9ci
15569555
3-fluoro-4-nitrobenzenesulfonyl chloride
[O-][N+](=O)C1=C(F)C=C(C=C1)S(Cl)(=O)=O
86156-93-6
C6H3ClFNO4S
239.60
MFCD03412240
AAWFUSFNPBRQDE-UHFFFAOYSA-N
3-fluoro-4-nitrobenzene-1-sulfonyl chloride, 3-fluoro-4-nitro-benzenesulfonyl chloride, 3-fluoro-4-nitrobenzenesulfonylchloride, 3-fluro-4-nitro-benzensulfonylchloride, 3-fluoro-4-nitrophenylsulfonyl chloride, benzenesulfonylchloride, 3-fluoro-4-nitro, benzenesulfonyl chloride, 3-fluoro-4-nitro, 3-fluoro-4-nitro-benzensulfonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-nitro-9ci
15569555
3-fluoro-4-nitrobenzenesulfonyl chloride
[O-][N+](=O)C1=C(F)C=C(C=C1)S(Cl)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 44°C to 48°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 239.61
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-nitrobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit