Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-4-nitrotoluene, 99%

Click to view available options
Quantity:
5g
25g
446-34-4
C7H6FNO2
155.13
MFCD00007053
WZMOWQCNPFDWPA-UHFFFAOYSA-N
3-fluoro-4-nitrotoluene, benzene, 2-fluoro-4-methyl-1-nitro, 2-fluoro-4-methylnitrobenzene, 2-fluoro-4-methyl-1-nitro-benzene, 3-fluoro-4-nitrobenzene, 2-fluor-4-methyl-1-nitrobenzol, pubchem1595, 3-fluoro4-nitrotoluene, acmc-1alaq, 3-fluoro-4-nitro toluene
67966
2-fluoro-4-methyl-1-nitrobenzene
CC1=CC=C(C(F)=C1)[N+]([O-])=O
446-34-4
C7H6FNO2
155.13
MFCD00007053
WZMOWQCNPFDWPA-UHFFFAOYSA-N
3-fluoro-4-nitrotoluene, benzene, 2-fluoro-4-methyl-1-nitro, 2-fluoro-4-methylnitrobenzene, 2-fluoro-4-methyl-1-nitro-benzene, 3-fluoro-4-nitrobenzene, 2-fluor-4-methyl-1-nitrobenzol, pubchem1595, 3-fluoro4-nitrotoluene, acmc-1alaq, 3-fluoro-4-nitro toluene
67966
2-fluoro-4-methyl-1-nitrobenzene
CC1=CC=C(C(F)=C1)[N+]([O-])=O
Flash Point >110°C (230°F)
Boiling Point 97°C to 98°C (3mmHg)
Density 1.438
Quantity 5g
Melting Point 51°C to 55°C
UN Number UN2811
Beilstein 2502584
Formula Weight 155.13
Percent Purity 99%
Chemical Name or Material 3-Fluoro-4-nitrotoluene

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 207-166-5

TSCA : Yes

Recommended Storage : Ambient temperatures