Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
211108-50-8
C10H16FNO3
217.24
JZNWQLLPLOQGOI-UHFFFAOYSA-N
1-boc-3-fluoro-4-oxopiperidine,3-fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester,1-boc-3-fluoro-4-piperidone,1-tert-butoxycarbonyl-3-fluoro-4-piperidone,1-piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester,1-tert-butoxycarbonyl-3-fluoro-4-oxopiperidine,3-fluoro-4-oxo-piperidine-1-carboxylic acid tert-butyl ester,tert-butyl3-fluoro-4-oxopiperidine-1-carboxylate,tert-butyl 3-fluoro-4-oxo-piperidine-1-carboxylate,n-boc-3-fluoro-4-piperidinone
10560711
tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(=O)C(C1)F
211108-50-8
C10H16FNO3
217.24
JZNWQLLPLOQGOI-UHFFFAOYSA-N
1-boc-3-fluoro-4-oxopiperidine,3-fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester,1-boc-3-fluoro-4-piperidone,1-tert-butoxycarbonyl-3-fluoro-4-piperidone,1-piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester,1-tert-butoxycarbonyl-3-fluoro-4-oxopiperidine,3-fluoro-4-oxo-piperidine-1-carboxylic acid tert-butyl ester,tert-butyl3-fluoro-4-oxopiperidine-1-carboxylate,tert-butyl 3-fluoro-4-oxo-piperidine-1-carboxylate,n-boc-3-fluoro-4-piperidinone
10560711
tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(=O)C(C1)F
Quantity 5g
Packaging Glass bottle
Melting Point 75°C to 77°C
Assay Percent Range 96% min. (GC)
Formula Weight 217.24
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning