Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-pyridinecarboxaldehyde, 97%, ACROS Organics

Click to view available options
Quantity:
1g
5g
40273-47-0
C6H4FNO
125.1
KLKQXQXSZCNWLZ-UHFFFAOYSA-N
3-fluoroisonicotinaldehyde,3-fluoropyridine-4-carboxaldehyde,3-fluoro-4-formylpyridine,3-fluoro-4-pyridinecarboxaldehyde,4-pyridinecarboxaldehyde, 3-fluoro,3-fluoro-pyridine-4-carbaldehyde,3-fluoro-4-pyridinecarbaldehyde,brettphospd,pubchem5137,acmc-20a1qb
2763003
3-fluoropyridine-4-carbaldehyde
C1=CN=CC(=C1C=O)F
40273-47-0
C6H4FNO
125.1
KLKQXQXSZCNWLZ-UHFFFAOYSA-N
3-fluoroisonicotinaldehyde,3-fluoropyridine-4-carboxaldehyde,3-fluoro-4-formylpyridine,3-fluoro-4-pyridinecarboxaldehyde,4-pyridinecarboxaldehyde, 3-fluoro,3-fluoro-pyridine-4-carbaldehyde,3-fluoro-4-pyridinecarbaldehyde,brettphospd,pubchem5137,acmc-20a1qb
2763003
3-fluoropyridine-4-carbaldehyde
C1=CN=CC(=C1C=O)F
Specific Gravity 1.255
Density 1.2550g/mL
Boiling Point 70.0°C (25.0 Torr)
Quantity 5g
Flash Point 67°C
Assay Percent Range 96% min. (GC)
Formula Weight 125.1
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-pyridinecarboxaldehyde

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning