Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3'-Fluoro-4'-(trifluoromethyl)acetophenone, 97+%

Click to view available options
Quantity:
1g
5g
237761-81-8
C9H6F4O
206.14
MFCD00236273
NXQBONPKAOTSNA-UHFFFAOYSA-N
3'-fluoro-4'-trifluoromethyl acetophenone,1-3-fluoro-4-trifluoromethyl phenyl ethanone,3-fluoro-4-trifluoromethyl acetophenone,1-3-fluoro-4-trifluoromethyl phenyl ethan-1-one,ethanone, 1-3-fluoro-4-trifluoromethyl phenyl,1-acetyl-3-fluoro-4-trifluoromethyl benzene,pubchem12023,acmc-1cij5,intermediates-zcf02639
2737532
1-[3-fluoro-4-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=CC(=C(C=C1)C(F)(F)F)F
237761-81-8
C9H6F4O
206.14
MFCD00236273
NXQBONPKAOTSNA-UHFFFAOYSA-N
3'-fluoro-4'-trifluoromethyl acetophenone,1-3-fluoro-4-trifluoromethyl phenyl ethanone,3-fluoro-4-trifluoromethyl acetophenone,1-3-fluoro-4-trifluoromethyl phenyl ethan-1-one,ethanone, 1-3-fluoro-4-trifluoromethyl phenyl,1-acetyl-3-fluoro-4-trifluoromethyl benzene,pubchem12023,acmc-1cij5,intermediates-zcf02639
2737532
1-[3-fluoro-4-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=CC(=C(C=C1)C(F)(F)F)F
Quantity 5g
Formula Weight 206.14
Percent Purity ≥97%
Chemical Name or Material 3'-Fluoro-4'-(trifluoromethyl)acetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning