Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-4-(trifluoromethyl)aniline, 97%

Click to view available options
Quantity:
1g
5g
69411-68-3
C7H5F4N
179.118
MFCD00190120
CRRVZRDISHOQQL-UHFFFAOYSA-N
4-amino-2-fluorobenzotrifluoride,3-fluoro-4-trifluoromethyl aniline,2-fluoro-4-amino benzotrifluoride,benzenamine, 3-fluoro-4-trifluoromethyl,3-fluoro-4-trifluoromethyl-phenylamine,3-fluoro-4-trifluoromethyl phenylamine,pubchem1631,4-amino,2-fluorobenzotrifluoride,3-fluoro-4-trifluoromethyl-aniline
2735914
3-fluoro-4-(trifluoromethyl)aniline
C1=CC(=C(C=C1N)F)C(F)(F)F
69411-68-3
C7H5F4N
179.118
MFCD00190120
CRRVZRDISHOQQL-UHFFFAOYSA-N
4-amino-2-fluorobenzotrifluoride,3-fluoro-4-trifluoromethyl aniline,2-fluoro-4-amino benzotrifluoride,benzenamine, 3-fluoro-4-trifluoromethyl,3-fluoro-4-trifluoromethyl-phenylamine,3-fluoro-4-trifluoromethyl phenylamine,pubchem1631,4-amino,2-fluorobenzotrifluoride,3-fluoro-4-trifluoromethyl-aniline
2735914
3-fluoro-4-(trifluoromethyl)aniline
C1=CC(=C(C=C1N)F)C(F)(F)F
Melting Point 54°C to 58°C
Quantity 1g
UN Number UN2811
Formula Weight 179.12
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)aniline

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.