Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-4-(trifluoromethyl)benzoic acid, 97+%

Click to view available options
Quantity:
1g
5g
115754-21-7
C8H4F4O2
208.112
MFCD00236279
HRIHSNPFVGMAKX-UHFFFAOYSA-N
3-fluoro-4-trifluoromethyl benzoic acid,benzoic acid, 3-fluoro-4-trifluoromethyl,alpha,alpha,alpha,3-tetrafluoro-p-toluic acid,pubchem4970,acmc-1c70e,ksc179k4t,3-fluoro-4 trifluoromethyl benzoic acid,3-fluoro-4-trifluoromethyl-benzoic acid,3-fluoro-4-trifluoromethyl benzoicacid,3-fluoro-4-trifluoromethyl benzoic aicd
688293
3-fluoro-4-(trifluoromethyl)benzoic acid
C1=CC(=C(C=C1C(=O)O)F)C(F)(F)F
115754-21-7
C8H4F4O2
208.112
MFCD00236279
HRIHSNPFVGMAKX-UHFFFAOYSA-N
3-fluoro-4-trifluoromethyl benzoic acid,benzoic acid, 3-fluoro-4-trifluoromethyl,alpha,alpha,alpha,3-tetrafluoro-p-toluic acid,pubchem4970,acmc-1c70e,ksc179k4t,3-fluoro-4 trifluoromethyl benzoic acid,3-fluoro-4-trifluoromethyl-benzoic acid,3-fluoro-4-trifluoromethyl benzoicacid,3-fluoro-4-trifluoromethyl benzoic aicd
688293
3-fluoro-4-(trifluoromethyl)benzoic acid
C1=CC(=C(C=C1C(=O)O)F)C(F)(F)F
Melting Point 173°C to 176°C
Quantity 1g
Formula Weight 208.11
Percent Purity ≥97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning