Alfa Aesar™ 3-Fluoro-4-(trifluoromethyl)benzonitrile, 97%

Manufacturer: Alfa Aesar™ B2379106

Quantity 5g

 View more versions of this product

Catalog No. AAB2379106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 231953-38-1
    Molecular Formula C8H3F4N
    Formula Weight 189.11
    MDL Number MFCD00236280
    UN Number UN3439
    Synonym 4-Cyano-2-fluorobenzotrifluoride; alpha,alpha,alpha,3-Tetrafluoro-p-tolunitrile
    Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzonitrile

    Specifications

    Formula Weight 189.11
    Molecular Formula C8H3F4N
    MDL Number MFCD00236280
    CAS 231953-38-1
    UN Number UN3439
    Synonym 4-Cyano-2-fluorobenzotrifluoride; alpha,alpha,alpha,3-Tetrafluoro-p-tolunitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Melting Point 45° to 48°C
    Quantity 5g
    Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzonitrile
    Assay 97%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit