Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
216144-68-2
C8H3ClF4O
226.555
MFCD00236284
DPDAEMNQJYDHRQ-UHFFFAOYSA-N
3-fluoro-4-trifluoromethyl benzoyl chloride,3-fluoro-4-trifluoromethyl benzoylchloride,3-fluoranyl-4-trifluoromethyl benzoyl chloride,acmc-1cobr,4-chlorocarbonyl-2-fluorobenzotrifluoride,3-fluoro-4-trifiuoromethyl benzoyl chloride,3-fluoro-4-trifluoromethyl benzoyl cloride,benzoylchloride, 3-fluoro-4-trifluoromethyl,benzoyl chloride, 3-fluoro-4-trifluoromethyl
2734709
3-fluoro-4-(trifluoromethyl)benzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)F)C(F)(F)F
216144-68-2
C8H3ClF4O
226.555
MFCD00236284
DPDAEMNQJYDHRQ-UHFFFAOYSA-N
3-fluoro-4-trifluoromethyl benzoyl chloride,3-fluoro-4-trifluoromethyl benzoylchloride,3-fluoranyl-4-trifluoromethyl benzoyl chloride,acmc-1cobr,4-chlorocarbonyl-2-fluorobenzotrifluoride,3-fluoro-4-trifiuoromethyl benzoyl chloride,3-fluoro-4-trifluoromethyl benzoyl cloride,benzoylchloride, 3-fluoro-4-trifluoromethyl,benzoyl chloride, 3-fluoro-4-trifluoromethyl
2734709
3-fluoro-4-(trifluoromethyl)benzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)F)C(F)(F)F
Packaging Glass bottle
Color Colorless to Yellow
Quantity 5g
Infrared Spectrum Authentic
Density 1.486g/mL
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H3(CF3)COCl
Formula Weight 226.56
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger