Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-4-(trifluoromethyl)benzylamine, 97%

Click to view available options
Quantity:
1g
5g
235106-09-9
C8H7F4N
193.145
MFCD00236287
XYWYEOHNURYGGP-UHFFFAOYSA-N
3-fluoro-4-trifluoromethyl benzylamine,3-fluoro-4-trifluoromethyl phenyl methanamine,3-fluoro-4-trifluoromethyl phenyl methylamine,1-3-fluoro-4-trifluoromethyl phenyl methanamine,acmc-1chwh,3-fluoro-4-trifluoromethylbenzylamine,3-fluoro-4-trifluoromethyl-benzylamine,benzenemethanamine,3-fluoro-4-trifluoromethyl
2737567
[3-fluoro-4-(trifluoromethyl)phenyl]methanamine
C1=CC(=C(C=C1CN)F)C(F)(F)F
235106-09-9
C8H7F4N
193.145
MFCD00236287
XYWYEOHNURYGGP-UHFFFAOYSA-N
3-fluoro-4-trifluoromethyl benzylamine,3-fluoro-4-trifluoromethyl phenyl methanamine,3-fluoro-4-trifluoromethyl phenyl methylamine,1-3-fluoro-4-trifluoromethyl phenyl methanamine,acmc-1chwh,3-fluoro-4-trifluoromethylbenzylamine,3-fluoro-4-trifluoromethyl-benzylamine,benzenemethanamine,3-fluoro-4-trifluoromethyl
2737567
[3-fluoro-4-(trifluoromethyl)phenyl]methanamine
C1=CC(=C(C=C1CN)F)C(F)(F)F
Quantity 1g
UN Number UN2735
Formula Weight 193.15
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzylamine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.