Alfa Aesar™ 3'-Fluoro-4'-(trifluoromethyl)propiophenone, 97+%

Manufacturer: Alfa Aesar™ B2281406

Quantity 5g

 View more versions of this product

Catalog No. AAB2281406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 237761-78-3
    Molecular Formula C10H8F4O
    Formula Weight 220.17
    MDL Number MFCD00236306
    Chemical Name or Material 3'-Fluoro-4'-(trifluoromethyl)propiophenone

    Specifications

    Formula Weight 220.17
    Quantity 5g
    Molecular Formula C10H8F4O
    MDL Number MFCD00236306
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point 35° to 38°C
    CAS 237761-78-3
    Chemical Name or Material 3'-Fluoro-4'-(trifluoromethyl)propiophenone
    Assay ≥97%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit