Alfa Aesar™ 3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 97%

Manufacturer: Alfa Aesar™ H5463403

Quantity 1g

 View more versions of this product

Catalog No. AAH5463403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1264616-18-3
    Molecular Formula C11H13BFNO3
    Formula Weight 237.04
    MDL Number MFCD20265305
    Beilstein 22139405
    Synonym 3-Fluoro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid
    Chemical Name or Material 3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid

    Specifications

    Formula Weight 237.04
    Molecular Formula C11H13BFNO3
    MDL Number MFCD20265305
    CAS 1264616-18-3
    Synonym 3-Fluoro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 1g
    Beilstein 22139405
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid
    Assay 97%

    Description

    Applications
    3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit