Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-5-iodopyridine, 95%

Click to view available options
Quantity:
250mg
1g
5g
757950-13-3
C5H3FIN
222.989
MFCD08457780
MMJSSIRVEYQWMU-UHFFFAOYSA-N
3-fluoro-5-iodo-pyridine,pyridine, 3-fluoro-5-iodo,3-fluoro-5-iodo-pyridin,pyridine,3-fluoro-5-iodo,3-fluoranyl-5-iodanyl-pyridine,3-iodo-5-fluoropyridine,5-fluoro-3-iodopyridine
18454778
3-fluoro-5-iodopyridine
C1=C(C=NC=C1I)F
757950-13-3
C5H3FIN
222.989
MFCD08457780
MMJSSIRVEYQWMU-UHFFFAOYSA-N
3-fluoro-5-iodo-pyridine,pyridine, 3-fluoro-5-iodo,3-fluoro-5-iodo-pyridin,pyridine,3-fluoro-5-iodo,3-fluoranyl-5-iodanyl-pyridine,3-iodo-5-fluoropyridine,5-fluoro-3-iodopyridine
18454778
3-fluoro-5-iodopyridine
C1=C(C=NC=C1I)F
Quantity 250mg
Color White
Sensitivity Light Sensitive
Beilstein 11107918
Formula Weight 222.99
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-Fluoro-5-iodopyridine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C