Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-5-methylbenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ H2664806

Quantity 5g

 View more versions of this product

Catalog No. AAH2664806


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

189628-39-5
138.14
5-Fluoro-m-tolualdehyde
C8H7FO
MFCD03094315
3-Fluoro-5-methylbenzaldehyde

Specifications

5g
C8H7FO
MFCD03094315
3-Fluoro-5-methylbenzaldehyde
189628-39-5
138.14
5-Fluoro-m-tolualdehyde
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit