Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5314903

Quantity 1g

 View more versions of this product

Catalog No. AAH5314903


  Check Availability
Add to cart

Specifications

Description

Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

871332-63-7
196.97
3-Fluoro-5-(methylaminocarbonyl)phenylboronic acid
C8H9BFNO3
MFCD07363793
3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid

Specifications

1g
C8H9BFNO3
MFCD07363793
3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid
871332-63-7
196.97
3-Fluoro-5-(methylaminocarbonyl)phenylboronic acid
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit