Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)aniline, 97%

Manufacturer: Alfa Aesar™ H2623806

Quantity 5g

 View more versions of this product

Catalog No. AAH2623806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 454-67-1
    Molecular Formula C7H5F4N
    Formula Weight 179.12
    MDL Number MFCD00061315
    UN Number UN2810
    Synonym 3-Amino-5-fluorobenzotrifluoride
    Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)aniline

    Specifications

    Formula Weight 179.12
    UN Number UN2810
    Odor Characteristic
    Molecular Formula C7H5F4N
    MDL Number MFCD00061315
    CAS 454-67-1
    Synonym 3-Amino-5-fluorobenzotrifluoride
    Health Hazard 2 GHS H Statement
    H311-H227-H302-H332-H315-H319
    Toxic in contact with skin.
    Combustible liquid.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Quantity 5g
    Health Hazard 1 Danger
    Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)aniline
    Assay 97%

    GHS H Statement
    H311-H227-H302-H332-H315-H319
    Toxic in contact with skin.
    Combustible liquid.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit