Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)anisole, 97%

Manufacturer: Alfa Aesar™ H2624003

Quantity 1g

 View more versions of this product

Catalog No. AAH2624003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 261951-79-5
    Molecular Formula C8H6F4O
    Formula Weight 194.13
    MDL Number MFCD01631491
    UN Number UN1993
    Synonym 3-Fluoro-5-methoxybenzotrifluoride
    Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)anisole

    Specifications

    Formula Weight 194.13
    Molecular Formula C8H6F4O
    MDL Number MFCD01631491
    CAS 261951-79-5
    UN Number UN1993
    Synonym 3-Fluoro-5-methoxybenzotrifluoride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 52°C (126°F)
    Quantity 1g
    Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)anisole
    Assay 97%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit