Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)benzyl bromide, 97%

Click to view available options
Quantity:
1g
5g
239087-09-3
C8H5BrF4
257.026
MFCD00061175
LCVWAXYGYMZJER-UHFFFAOYSA-N
3-fluoro-5-trifluoromethyl benzyl bromide,1-bromomethyl-3-fluoro-5-trifluoromethyl benzene,3-fluoro-5-trifluoromethylbenzyl bromide,3-fluoro-5-trifluoromethyl benzylbromide,benzene, 1-bromomethyl-3-fluoro-5-trifluoromethyl,acmc-1cdjv,3-trifluoromethyl-5-fluorobenzyl bromide,3-fluoro-5-trifluoromethyl-benzyl bromide,5-bromomethyl-3-fluoro-1-trifluoromethyl benzene
2737575
1-(bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene
C1=C(C=C(C=C1C(F)(F)F)F)CBr
239087-09-3
C8H5BrF4
257.026
MFCD00061175
LCVWAXYGYMZJER-UHFFFAOYSA-N
3-fluoro-5-trifluoromethyl benzyl bromide,1-bromomethyl-3-fluoro-5-trifluoromethyl benzene,3-fluoro-5-trifluoromethylbenzyl bromide,3-fluoro-5-trifluoromethyl benzylbromide,benzene, 1-bromomethyl-3-fluoro-5-trifluoromethyl,acmc-1cdjv,3-trifluoromethyl-5-fluorobenzyl bromide,3-fluoro-5-trifluoromethyl-benzyl bromide,5-bromomethyl-3-fluoro-1-trifluoromethyl benzene
2737575
1-(bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene
C1=C(C=C(C=C1C(F)(F)F)F)CBr
Boiling Point 43°C (1mmHg)
Quantity 5g
UN Number UN1760
Formula Weight 257.03
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.