Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)phenol, 97+%

Click to view available options
Quantity:
2g
10g
172333-87-8
C7H4F4O
180.102
MFCD00061288
VNTKPYNBATZMSV-UHFFFAOYSA-N
3-fluoro-5-trifluoromethyl phenol,5-fluoro-3-trifluoromethyl phenol,phenol, 3-fluoro-5-trifluoromethyl,3-fluoranyl-5-trifluoromethyl phenol,acmc-1c2u3,3-fluoro-5-trifluoromethyl-phenol,phenol,3-fluoro-5-trifluoromethyl,5-fluoro-alpha,alpha,alpha-trifluoro-m-cresol
2737587
3-fluoro-5-(trifluoromethyl)phenol
C1=C(C=C(C=C1O)F)C(F)(F)F
172333-87-8
C7H4F4O
180.102
MFCD00061288
VNTKPYNBATZMSV-UHFFFAOYSA-N
3-fluoro-5-trifluoromethyl phenol,5-fluoro-3-trifluoromethyl phenol,phenol, 3-fluoro-5-trifluoromethyl,3-fluoranyl-5-trifluoromethyl phenol,acmc-1c2u3,3-fluoro-5-trifluoromethyl-phenol,phenol,3-fluoro-5-trifluoromethyl,5-fluoro-alpha,alpha,alpha-trifluoro-m-cresol
2737587
3-fluoro-5-(trifluoromethyl)phenol
C1=C(C=C(C=C1O)F)C(F)(F)F
Quantity 2g
UN Number UN3265
Formula Weight 180.1
Percent Purity ≥97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)phenol

GHS H Statement
H301-H314-H318-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.