Alfa Aesar™ 3-Fluoro-DL-phenylglycine, 97+%

Manufacturer: Alfa Aesar™ B2281209

Quantity 10g

 View more versions of this product

Catalog No. AAB2281209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7292-74-2
    Molecular Formula C8H8FNO2
    Formula Weight 169.16
    MDL Number MFCD00049325
    Chemical Name or Material 3-Fluoro-DL-phenylglycine

    Specifications

    Formula Weight 169.16
    Molecular Formula C8H8FNO2
    MDL Number MFCD00049325
    CAS 7292-74-2
    Melting Point ≈290°C (subl.)
    Quantity 10g
    Chemical Name or Material 3-Fluoro-DL-phenylglycine
    Assay ≥97%

    Description

    Applications
    3-Fluoro-DL-phenylglycine is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit