Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Fluoro-N-methylaniline, 97%

Click to view available options
Quantity:
10g
50g
250g
1978-37-6
C7H8FN
125.146
MFCD02683095
FHYDHJXZZQCXOX-UHFFFAOYSA-N
n-methyl-3-fluoroaniline,3-fluoro-n-methylbenzenamine,3-fluoro-n-methylanilin,n-methyl-m-fluoroaniline,acmc-209xf6,ksc497i4f,3-fluoro-phenyl-methyl-amine,3-fluoro-n-methylaniline,benzenamine, 3-fluoro-n-methyl
2759011
3-fluoro-N-methylaniline
CNC1=CC(=CC=C1)F
1978-37-6
C7H8FN
125.146
MFCD02683095
FHYDHJXZZQCXOX-UHFFFAOYSA-N
n-methyl-3-fluoroaniline,3-fluoro-n-methylbenzenamine,3-fluoro-n-methylanilin,n-methyl-m-fluoroaniline,acmc-209xf6,ksc497i4f,3-fluoro-phenyl-methyl-amine,3-fluoro-n-methylaniline,benzenamine, 3-fluoro-n-methyl
2759011
3-fluoro-N-methylaniline
CNC1=CC(=CC=C1)F
Boiling Point 94°C (19mmHg)
Flash Point 86°C (187°F)
Odor Characteristic
Quantity 250g
UN Number UN2810
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 3-Fluoro-N-methylaniline

GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P260-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.