Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3'-Fluoroacetanilide, 98%

Click to view available options
Quantity:
5g
25g
100g
351-28-0
C8H8FNO
153.156
MFCD00017917
AQLLDCFUQXGLHM-UHFFFAOYSA-N
3'-fluoroacetanilide,n-3-fluorophenyl acetamide,3-fluoroacetanilide,m-fluoroacetanilide,3-acetamido-1-fluorobenzene,acetanilide, 3'-fluoro,1-fluoro-3-acetamidobenzene,acetamide, n-3-fluorophenyl,acmc-209idd,n-acetyl-3-fluoroaniline
9594
N-(3-fluorophenyl)acetamide
CC(=O)NC1=CC(=CC=C1)F
351-28-0
C8H8FNO
153.156
MFCD00017917
AQLLDCFUQXGLHM-UHFFFAOYSA-N
3'-fluoroacetanilide,n-3-fluorophenyl acetamide,3-fluoroacetanilide,m-fluoroacetanilide,3-acetamido-1-fluorobenzene,acetanilide, 3'-fluoro,1-fluoro-3-acetamidobenzene,acetamide, n-3-fluorophenyl,acmc-209idd,n-acetyl-3-fluoroaniline
9594
N-(3-fluorophenyl)acetamide
CC(=O)NC1=CC(=CC=C1)F
Melting Point 86°C to 89°C
Quantity 5g
Beilstein 2803073
Formula Weight 153.16
Percent Purity 98%
Chemical Name or Material 3'-Fluoroacetanilide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning