Alfa Aesar™ 3'-Fluoroacetophenone, 98+%

Manufacturer: Alfa Aesar™ A1173322

Quantity 100g

 View more versions of this product

Catalog No. AAA1173322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 455-36-7
    Molecular Formula C8H7FO
    Formula Weight 138.14
    MDL Number MFCD00000338
    Beilstein 907041
    Chemical Name or Material 3'-Fluoroacetophenone

    Specifications

    Formula Weight 138.14
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    Quantity 100g
    MDL Number MFCD00000338
    CAS 455-36-7
    Melting Point −3°C
    Molecular Formula C8H7FO
    Beilstein 907041
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Flash Point 80°C (176°F)
    Boiling Point 80° to 81°C (9mmHg)
    Chemical Name or Material 3'-Fluoroacetophenone
    Assay ≥98%

    Description

    Applications
    3 ft.-Fluoroacetophenone is used as raw material and intermediate in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

    Solubility
    Slightly soluble in water.

    Notes
    Incompatible with strong bases, strong oxidizing agents, strong reducing agents. Ensure adequate ventilation during use. Use only in a chemical fume hood.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit