Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Fluoroacetophenone, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
455-36-7
C8H7FO
138.14
MFCD00000338
HCEKGPAHZCYRBZ-UHFFFAOYSA-N
3'-fluoroacetophenone,1-3-fluorophenyl ethanone,3-fluoroacetophenone,m-fluoroacetophenone,ethanone, 1-3-fluorophenyl,1-3-fluorophenyl ethan-1-one,acetophenone, 3'-fluoro,3'-fluoro-acetophenone,1-acetyl-3-fluorobenzene
9967
1-(3-fluorophenyl)ethanone
CC(=O)C1=CC(=CC=C1)F
455-36-7
C8H7FO
138.14
MFCD00000338
HCEKGPAHZCYRBZ-UHFFFAOYSA-N
3'-fluoroacetophenone,1-3-fluorophenyl ethanone,3-fluoroacetophenone,m-fluoroacetophenone,ethanone, 1-3-fluorophenyl,1-3-fluorophenyl ethan-1-one,acetophenone, 3'-fluoro,3'-fluoro-acetophenone,1-acetyl-3-fluorobenzene
9967
1-(3-fluorophenyl)ethanone
CC(=O)C1=CC(=CC=C1)F
Specific Gravity 1.12
Density 1.1200g/mL
Boiling Point 81.0°C (9.0 mmHg)
Quantity 1g
Refractive Index 1.5080 to 1.5100
Packaging Glass bottle
Flash Point 80°C
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula FC6H4COCH3
Beilstein 07, III, 961
Formula Weight 138.14
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3′-Fluoroacetophenone
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 207-245-4