Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoroaniline 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
372-19-0
C6H6FN
111.12
MFCD00007758
QZVQQUVWFIZUBQ-UHFFFAOYSA-N
m-fluoroaniline,1-amino-3-fluorobenzene,3-fluoranilin,benzenamine, 3-fluoro,3-fluorobenzenamine,3-fluoro-phenylamine,aniline, 3-fluoro,3-fluorophenylamine,aniline, m-fluoro,3-fluoranilin czech
9742
CHEBI:27873
3-fluoroaniline
C1=CC(=CC(=C1)F)N
372-19-0
C6H6FN
111.12
MFCD00007758
QZVQQUVWFIZUBQ-UHFFFAOYSA-N
m-fluoroaniline,1-amino-3-fluorobenzene,3-fluoranilin,benzenamine, 3-fluoro,3-fluorobenzenamine,3-fluoro-phenylamine,aniline, 3-fluoro,3-fluorophenylamine,aniline, m-fluoro,3-fluoranilin czech
9742
CHEBI:27873
3-fluoroaniline
C1=CC(=CC(=C1)F)N
Specific Gravity 1.15
Flash Point 77°C
Boiling Point 186.0°C (756.0 mmHg)
Color Yellow to Brown
Quantity 500mL
Density 1.1500g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5430 to 1.5450
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula FC6H4NH2
Beilstein 12,597
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in carbon tetrachloride,diethyl ether,,ethanol
Formula Weight 111.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Fluoroaniline, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-747-0

RTECSNumber : BY1400000

TSCA : TSCA