Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoroanisole, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
456-49-5
C7H7FO
126.13
MFJNOXOAIFNSBX-UHFFFAOYSA-N
3-fluoroanisole,m-fluoroanisole,benzene, 1-fluoro-3-methoxy,1-fluoro-3-methoxy-benzene,2-fluoro-4-methoxybenzene,fr co1 wln,3-fluoroanisol,3-fluoro-anisole,3-fluoro anisole,anisole, m-fluoro
9975
1-fluoro-3-methoxybenzene
COC1=CC(=CC=C1)F
456-49-5
C7H7FO
126.13
MFJNOXOAIFNSBX-UHFFFAOYSA-N
3-fluoroanisole,m-fluoroanisole,benzene, 1-fluoro-3-methoxy,1-fluoro-3-methoxy-benzene,2-fluoro-4-methoxybenzene,fr co1 wln,3-fluoroanisol,3-fluoro-anisole,3-fluoro anisole,anisole, m-fluoro
9975
1-fluoro-3-methoxybenzene
COC1=CC(=CC=C1)F

May darken on storage

Melting Point -35.0°C
Specific Gravity 1.104
Density 1.1040g/mL
Flash Point 43°C
Refractive Index 1.4876 to 1.4896
Boiling Point 158.0°C (743.0 mmHg)
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H4OCH3
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 3-Fluoroanisole
Show More Show Less

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.

GHS Signal Word: Warning

EINECSNumber : 207-267-4